3-(4-Bromophenyl)-1H-[1,2,4]triazole ≥95%

Supplier: Matrix Scientific

073934-250MG 073934-1G
102533-778EA 684.62 USD
102533-778 102533-780
3-(4-Bromophenyl)-1H-[1,2,4]triazole ≥95%
3-(4-Bromophenyl)-1H-[1,2,4]triazole

Hazard: Irritant

Formula: C₈H₆BrN₃
MW: 224.0600 g/mol
MDL Number: MFCD16659605
CAS Number: 118863-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR