1-(2,6-Dichloro-3-fluorophenyl)ethanol ≥95%

Supplier: Matrix Scientific

073933-1G 073933-5G 073933-10G
102533-772EA 427.69 USD
102533-772 102533-774 102533-776
1-(2,6-Dichloro-3-fluorophenyl)ethanol ≥95%
1-(2,6-Dichloro-3-fluorophenyl)ethanol

Hazard: Irritant

Formula: C₈H₇Cl₂FO
MW: 209.0450 g/mol
MDL Number: MFCD08443334
CAS Number: 756520-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR