2,4-Dichloro-7-methylthieno[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

073932-1G 073932-5G
102533-768EA 533.85 USD
102533-768 102533-770
2,4-Dichloro-7-methylthieno[3,2-d]pyrimidine ≥95%
2,4-Dichloro-7-methylthieno[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₇H₄Cl₂N₂S
MW: 219.0900 g/mol
MDL Number: MFCD08061253
CAS Number: 35265-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR