1,14-Dibromo-3,6,9,12-tetraoxatetradecane ≥95%

Supplier: Matrix Scientific

Synonyms: Bromo-PEG4-bromide

073931-250MG 073931-1G
102533-764EA 826.15 USD
102533-764 102533-766
1,14-Dibromo-3,6,9,12-tetraoxatetradecane ≥95%
1,14-Dibromo-3,6,9,12-tetraoxatetradecane

Hazard: Irritant

Formula: C₁₀H₂₀Br₂O₄
MW: 364.071 g/mol
MDL Number: MFCD07369317
CAS Number: 57602-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR