(R)-4-Boc-(3-hydroxymethyl)morpholine ≥95%

Supplier: Matrix Scientific

073929-1G 073929-5G
102533-754EA 541.54 USD
102533-754 102533-756
(R)-4-Boc-(3-hydroxymethyl)morpholine ≥95%
(R)-4-Boc-(3-hydroxymethyl)morpholine

Hazard: Irritant

Formula: C₁₀H₁₉NO₄
MDL Number: MFCD06799477
CAS Number: 215917-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR