1-(Pyrimidin-5-yl)ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Acetylpyrimidine

073927-1G 073927-5G
102533-742EA 266.15 USD
102533-742 102533-744
1-(Pyrimidin-5-yl)ethanone ≥95%
1-(Pyrimidin-5-yl)ethanone

Hazard: Irritant

Formula: C₆H₆N₂O
MW: 122.1246 g/mol
MDL Number: MFCD09038477
CAS Number: 10325-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR