5-Bromobenzofuran ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromobenzo[b]furan

073926-1G 073926-5G 073926-10G
102533-736EA 178.46 USD
102533-736 102533-738 102533-740
5-Bromobenzofuran ≥95%
5-Bromobenzofuran

Hazard: Irritant

Formula: C₈H₅BrO
MW: 197.04 g/mol
MDL Number: MFCD03407317
CAS Number: 23145-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR