7-Bromo-4-chloro-3-nitroquinoline ≥95%
Supplier: Matrix Scientific
073924-250MG
073924-1G
102533-726EA
967.69
USD
102533-726
102533-728
7-Bromo-4-chloro-3-nitroquinoline ≥95%
7-Bromo-4-chloro-3-nitroquinoline
Hazard: Irritant
Formula:
C₉H₄BrClN₂O₂ MW: 287.5000 g/mol |
MDL Number:
MFCD12828220 CAS Number: 723280-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...