4-Phenylpyridin-2-ol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Hydroxy-4-phenylpyridine
073923-250MG
073923-1G
073923-5G
102533-720EA
640
USD
102533-720
102533-722
102533-724
4-Phenylpyridin-2-ol ≥95%
4-Phenylpyridin-2-ol
Hazard: Irritant
Formula:
C₁₁H₉NO MW: 171.1953 g/mol |
MDL Number:
MFCD07437954 CAS Number: 19006-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...