4-Chloro-6-iodoquinazoline ≥95%

Supplier: Matrix Scientific

073921-1G 073921-5G
102533-710EA 427.69 USD
102533-710 102533-712
4-Chloro-6-iodoquinazoline ≥95%
4-Chloro-6-iodoquinazoline

Hazard: Irritant

Formula: C₈H₄ClIN₂
MW: 290.4900 g/mol
MDL Number: MFCD01862193
CAS Number: 98556-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR