4-(Aminomethyl)benzenesulfonamide hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
alpha-Amino-p-toluenesulphonamide hydrochloride, Homosulphamine hydrochloride, Mafenide hydrochloride, Mafenide HCl
073920-25G
073920-100G
102663-960EA
75.38
USD
102663-960
102663-962
4-(Aminomethyl)benzenesulfonamide hydrochloride ≥95%
4-(Aminomethyl)benzenesulfonamide hydrochloride
Hazard: Irritant/Refrigerate
Formula:
C₇H₁₁ClN₂O₂S MW: 222.6924 g/mol |
MDL Number:
MFCD00013005 CAS Number: 138-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...