2-(Methoxymethyl)pyridine ≥95%

Supplier: Matrix Scientific

073919-1G 073919-5G 073919-10G
102533-700EA 355.38 USD
102533-700 102533-702 102533-704
2-(Methoxymethyl)pyridine ≥95%
2-(Methoxymethyl)pyridine

Hazard: Irritant

Formula: C₇H₉NO
MW: 123.1500 g/mol
MDL Number: MFCD00030825
CAS Number: 23579-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR