1-Boc-4-Cbz-aminoazepane ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(((benzyloxy)carbonyl)amino)azepane-1-carboxylate

073918-250MG 073918-1G
102533-696EA 967.69 USD
102533-696 102533-698
1-Boc-4-Cbz-aminoazepane ≥95%
1-Boc-4-Cbz-aminoazepane

Hazard: Irritant

Formula: C₁₉H₂₈N₂O₄
MW: 348.436 g/mol
MDL Number: MFCD18207774
CAS Number: 878630-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR