Methyl-5-amino-1H-1,2,4-triazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

073914-1G 073914-5G
102533-674EA 427.69 USD
102533-674 102533-676
Methyl-5-amino-1H-1,2,4-triazole-3-carboxylate ≥95%
Methyl-5-amino-1H-1,2,4-triazole-3-carboxylate

Hazard: Irritant

Formula: C₄H₆N₄O₂
MDL Number: MFCD00297193
CAS Number: 3641-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR