5-Chloropyrimidine-2-carbonitrile ≥95%

Supplier: Matrix Scientific

073912-250MG 073912-1G 073912-5G
102533-662EA 1018.46 USD
102533-662 102533-664 102533-666
5-Chloropyrimidine-2-carbonitrile ≥95%
5-Chloropyrimidine-2-carbonitrile

Hazard: Irritant

Formula: C5H2ClN3
MW: 139.5400 g/mol
MDL Number: MFCD03001466
CAS Number: 38275-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR