5-(2-Hydroxyethylamino)-2-methoxylaniline sulfate ≥95%

Supplier: Matrix Scientific

073911-1G 073911-5G
102533-658EA 112.31 USD
102533-658 102533-660
5-(2-Hydroxyethylamino)-2-methoxylaniline sulfate ≥95%
5-(2-Hydroxyethylamino)-2-methoxylaniline sulfate

Hazard: Irritant/Refrigerate

Formula: C₉H₁₆N₂O₆S
MW: 280.3000 g/mol
MDL Number: MFCD00467095
CAS Number: 83763-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR