(S)-Tetrahydrofuran-3-amine hydrochloride ≥95%

Supplier: Matrix Scientific

073909-250MG 073909-1G 073909-5G
102533-648EA 561.54 USD
102533-648 102533-650 102533-652
(S)-Tetrahydrofuran-3-amine hydrochloride ≥95%
(S)-Tetrahydrofuran-3-amine hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD08445642
CAS Number: 204512-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR