3,4,5-Trichloropyridazine ≥95%
Supplier: Matrix Scientific
073907-1G
073907-5G
102533-636EA
44.62
USD
102533-636
102533-638
3,4,5-Trichloropyridazine ≥95%
3,4,5-Trichloropyridazine
Hazard: Toxic
Formula:
C₄HCl₃N₂ MW: 183.42 Melting Pt: 60…62 °C |
MDL Number:
MFCD00006464 CAS Number: 14161-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...