(3,5-Dichlorophenyl)acetic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(3,5-Dichlorophenyl)acetic acid
073904-5G
073904-25G
102533-624EA
110.77
USD
102533-624
102551-856
(3,5-Dichlorophenyl)acetic acid ≥95%
(3,5-Dichlorophenyl)acetic acid
Hazard: Irritant
Formula:
C₈H₆Cl₂O₂ MW: 205.03 g/mol |
MDL Number:
MFCD01764671 CAS Number: 51719-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...