3,4'-Dichlorodiphenyl ether ≥95%
Supplier: Matrix Scientific
073902-1G
073902-5G
102533-612EA
533.85
USD
102533-612
102533-614
3,4'-Dichlorodiphenyl ether ≥95%
3,4'-Dichlorodiphenyl ether
Hazard: Irritant
Formula:
C12H8Cl2O MW: 239.1 g/mol |
MDL Number:
MFCD00043876 CAS Number: 6842-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...