2-Chloroethyldimethylammonium chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
N-(2-Chloroethyl)-N,N-Dimethylammonium Chloride, 2-Dimethylaminoethyl chloride hydrochloride
073901-5G
073901-10G
073901-100G
102533-606EA
36.92
USD
102533-606
102533-608
102533-610
2-Chloroethyldimethylammonium chloride ≥95%
2-Chloroethyldimethylammonium chloride
Hazard: Toxic/Hygroscopic
Formula:
C₄H₁₁Cl₂N MW: 144.04 g/mol Melting Pt: 205-208°C |
MDL Number:
MFCD00012516 CAS Number: 4584-46-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...