4-Bromo-1H-benzoimidazole ≥95%

Supplier: Matrix Scientific

073900-250MG 073900-1G 073900-5G
102533-600EA 189.23 USD
102533-600 102533-602 102533-604
4-Bromo-1H-benzoimidazole ≥95%
4-Bromo-1H-benzoimidazole

Hazard: Irritant/Store in Freezer

Formula: C₇H₅BrN₂
MW: 197.0320 g/mol
MDL Number: MFCD09754164
CAS Number: 83741-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR