N-(2-Bromoethyl)-1,3-propanediamine dihydrobromide ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(2-Bromoethylamino)propylamine dihydrobromide

073899-1G 073899-5G 073899-10G
102533-594EA 107.69 USD
102533-594 102533-596 102533-598
N-(2-Bromoethyl)-1,3-propanediamine dihydrobromide ≥95%
N-(2-Bromoethyl)-1,3-propanediamine dihydrobromide

Hazard: Irritant

Formula: C₅H₁₅Br₃N₂
MW: 342.90 g/mol
MDL Number: MFCD00460336
CAS Number: 23545-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR