3-Amino-5-methylbenzotrifluoride ≥95%

Supplier: Matrix Scientific

073898-1G 073898-5G
102533-590EA 284.62 USD
102533-590 102533-592
3-Amino-5-methylbenzotrifluoride ≥95%
3-Amino-5-methylbenzotrifluoride

Hazard: Irritant

Formula: C₈H₈F₃N
MW: 175.1510 g/mol
MDL Number: MFCD01631580
CAS Number: 96100-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR