2,2'-Bipyridine-5,5'-dicarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
6,6′-Dinicotinic acid
073897-1G
073897-5G
102533-586EA
284.62
USD
102533-586
102533-588
2,2'-Bipyridine-5,5'-dicarboxylic acid ≥95%
2,2'-Bipyridine-5,5'-dicarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₈N₂O₄ MW: 244.21 g/mol |
MDL Number:
MFCD01318320 CAS Number: 1802-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...