2-(4-Fluorophenyl)propanoic acid ≥95%

Supplier: Matrix Scientific

073896-1G 073896-5G 073896-10G
102533-580EA 427.69 USD
102533-580 102533-582 102533-584
2-(4-Fluorophenyl)propanoic acid ≥95%
2-(4-Fluorophenyl)propanoic acid

Hazard: Irritant

Formula: C₉H₉FO₂
MDL Number: MFCD00054492
CAS Number: 75908-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR