(R)-(-)-2-Methylpiperazine ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-2-Methylpiperazine

073895-5G 073895-100G
102533-574EA 78.46 USD
102533-574 102533-578
(R)-(-)-2-Methylpiperazine ≥95%
(R)-(-)-2-Methylpiperazine

Hazard: Irritant

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 153
MDL Number: MFCD00192317
CAS Number: 75336-86-6
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR