2,4-Dihydroxy-3-methylbenzaldehyde ≥95%

Supplier: Matrix Scientific

073893-1G 073893-5G
102533-562EA 53.85 USD
102533-562 102533-564
2,4-Dihydroxy-3-methylbenzaldehyde ≥95%
2,4-Dihydroxy-3-methylbenzaldehyde

Hazard: Irritant

Formula: C₈H₈O₃
MW: 152.1500 g/mol
MDL Number: MFCD00156883
CAS Number: 6248-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR