3-(1-Cyanoethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

073890-10G 073890-100G
102533-546EA 284.62 USD
102533-546 102533-548
3-(1-Cyanoethyl)benzoic acid ≥95%
3-(1-Cyanoethyl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00002520
CAS Number: 5537-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR