1-(5-Chloro-1H-indazol-1-yl)ethanone ≥95%

Supplier: Matrix Scientific

073886-1G 073886-5G 073886-10G
102533-524EA 233.85 USD
102533-524 102533-526 102533-528
1-(5-Chloro-1H-indazol-1-yl)ethanone ≥95%
1-(5-Chloro-1H-indazol-1-yl)ethanone

Hazard: Irritant

Formula: C₉H₇ClN₂O
MW: 194.6177 g/mol
MDL Number: MFCD11109546
CAS Number: 98083-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR