(4-Bromo-2-fluorophenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-fluorobenzyl cyanide, 2-(4-Bromo-2-fluorophenyl)acetonitrile

073884-5G 073884-10G 073884-100G
102533-512EA 61.54 USD
102533-512 102533-514 102533-516
(4-Bromo-2-fluorophenyl)acetonitrile ≥95%
(4-Bromo-2-fluorophenyl)acetonitrile

Hazard: Toxic

Formula: C₈H₅BrFN
MW: 214.0300 g/mol
Melting Pt: 52…53 °C
MDL Number: MFCD07368653
CAS Number: 114897-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR