2,4-Dichloro-3-fluorobenzonitrile ≥95%
Supplier: Matrix Scientific
073880-1G
073880-5G
073880-10G
102533-488EA
284.62
USD
102533-488
102533-490
102533-492
2,4-Dichloro-3-fluorobenzonitrile ≥95%
2,4-Dichloro-3-fluorobenzonitrile
Hazard: Irritant
Formula:
C₇H₂Cl₂FN MW: 190.0000 g/mol |
MDL Number:
MFCD06658268 CAS Number: 161612-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...