2-Bromoquinoline-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

073878-250MG 073878-1G 073878-5G
102533-478EA 355.38 USD
102533-478 102533-480 102533-482
2-Bromoquinoline-4-carboxylic acid ≥95%
2-Bromoquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆BrNO₂
MW: 252.0600 g/mol
MDL Number: MFCD08705669
CAS Number: 15733-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR