(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane ≥95%

Supplier: Matrix Scientific

Synonyms: tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

073876-1G 073876-5G
102533-468EA 801.54 USD
102533-468 102533-470
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane ≥95%
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane

Hazard: Irritant

Formula: C₁₀H₁₈N₂O₂
MW: 198.26 g/mol
MDL Number: MFCD01569250
CAS Number: 113451-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR