2-Amino-3-(methoxycarbonyl)benzoic acid ≥95%

Supplier: Matrix Scientific

073875-1G 073875-5G
102533-464EA 300 USD
102533-464 102533-466
2-Amino-3-(methoxycarbonyl)benzoic acid ≥95%
2-Amino-3-(methoxycarbonyl)benzoic acid

Hazard: Irritant

Formula: C₉H₉NO₄
MW: 195.1721 g/mol
MDL Number: MFCD06200897
CAS Number: 253120-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR