Methyl-4-bromo-3-methylbenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-bromo-3-methylbenzoate
073874-10G
073874-100G
102533-460EA
143.08
USD
102533-460
102533-462
Methyl-4-bromo-3-methylbenzoate ≥95%
Methyl 4-bromo-3-methylbenzoate
Hazard: Irritant
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol |
MDL Number:
MFCD00673014 CAS Number: 148547-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...