4-Iodo-3-nitrobenzamide ≥95%

Supplier: Matrix Scientific

Synonyms: Iniparib

073871-1G 073871-5G
102533-446EA 640 USD
102533-446 102533-448
4-Iodo-3-nitrobenzamide ≥95%
4-Iodo-3-nitrobenzamide

Hazard: Irritant

Formula: C₇H₅IN₂O₃
MW: 292.0307 g/mol
MDL Number: MFCD11110639
CAS Number: 160003-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR