7-Chloro-4-quinazolone ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Chloro-4-hydroxyquinazoline

073870-1G 073870-5G 073870-10G
102533-440EA 23.08 USD
102533-440 102533-442 102533-444
7-Chloro-4-quinazolone ≥95%
7-Chloro-4-quinazolone

Hazard: Irritant

Formula: C₈H₅ClN₂O
MW: 180.5900 g/mol
MDL Number: MFCD01570890
CAS Number: 31374-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR