5-Bromopyrazolo[1,5-a]pyrimidine ≥95%

Supplier: Matrix Scientific

073868-250MG 073868-1G
102533-430EA 684.62 USD
102533-430 102533-432
5-Bromopyrazolo[1,5-a]pyrimidine ≥95%
5-Bromopyrazolo[1,5-a]pyrimidine

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
MDL Number: MFCD11042744
CAS Number: 1159981-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR