2-Hydroxy-4-methylthiazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

073866-250MG 073866-1G 073866-5G
102533-418EA 355.38 USD
102533-418 102533-420 102533-422
2-Hydroxy-4-methylthiazole-5-carboxylic acid ≥95%
2-Hydroxy-4-methylthiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₅H₅NO₃S
MW: 159.1600 g/mol
MDL Number: MFCD14706171
CAS Number: 875237-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR