3-Methyl-5-nitro-1H-pyrazole ≥95%

Supplier: Matrix Scientific

073865-1G 073865-5G 073865-10G
102533-412EA 123.08 USD
102533-412 102533-414 102533-416
3-Methyl-5-nitro-1H-pyrazole ≥95%
3-Methyl-5-nitro-1H-pyrazole

Hazard: Irritant

Formula: C4H5N3O2
MW: 127.1014 g/mol
MDL Number: MFCD00463977
CAS Number: 34334-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR