4,4-Dimethylcyclohexanol ≥95%

Supplier: Matrix Scientific

073861-1G 073861-5G
102533-390EA 123.08 USD
102533-390 102533-392
4,4-Dimethylcyclohexanol ≥95%
4,4-Dimethylcyclohexanol

Hazard: Irritant

Formula: C₈H₁₆O
MW: 128.2100 g/mol
MDL Number: MFCD00101954
CAS Number: 932-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR