(4-Bromo-2-methoxyphenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

073855-250MG 073855-1G 073855-5G
102533-356EA 2873.85 USD
102533-356 102533-358 102533-360
(4-Bromo-2-methoxyphenyl)acetonitrile ≥95%
(4-Bromo-2-methoxyphenyl)acetonitrile

Hazard: Irritant

Formula: C₉H₈BrNO
MW: 226.0699 g/mol
MDL Number: MFCD11847504
CAS Number: 858523-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR