3-Aminopyrazolo[1,5-a]pyrimidine ≥95%

Supplier: Matrix Scientific

073854-250MG 073854-1G 073854-5G
102533-350EA 2850.77 USD
102533-350 102533-352 102533-354
3-Aminopyrazolo[1,5-a]pyrimidine ≥95%
3-Aminopyrazolo[1,5-a]pyrimidine

Hazard: Irritant

Formula: C6H6N4
MW: 134.1400 g/mol
MDL Number: MFCD00501923
CAS Number: 232600-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR