3-Chloro-5-nitropyridin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3-chloro-5-nitropyridine

073853-250MG 073853-1G 073853-5G
102533-344EA 924.62 USD
102533-344 102533-346 102533-348
3-Chloro-5-nitropyridin-4-amine ≥95%
3-Chloro-5-nitropyridin-4-amine

Hazard: Irritant

Formula: C5H4ClN3O2
MW: 173.5572 g/mol
MDL Number: MFCD14706168
CAS Number: 89284-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR