2,6-Dichloro-4-(piperidin-1-ylcarbonyl)pyridine ≥95%

Supplier: Matrix Scientific

073852-250MG 073852-1G 073852-5G
102533-338EA 498.46 USD
102533-338 102533-340 102533-342
2,6-Dichloro-4-(piperidin-1-ylcarbonyl)pyridine ≥95%
2,6-Dichloro-4-(piperidin-1-ylcarbonyl)pyridine

Hazard: Irritant

Formula: C₁₁H₁₂Cl₂N₂O
MW: 259.13 g/mol
MDL Number: MFCD01764510
CAS Number: 287196-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR