1-Quinolin-5-ylmethylamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Quinolin-5-yl-methylamine hydrochloride

073850-250MG 073850-1G 073850-5G
102533-326EA 498.46 USD
102533-326 102533-328 102533-330
1-Quinolin-5-ylmethylamine hydrochloride ≥95%
1-Quinolin-5-ylmethylamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂
MW: 194.6600 g/mol
MDL Number: MFCD09701405
CAS Number: 1187931-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR