1-(Benzo[d]oxazol-2-yl)ethanone ≥95%

Supplier: Matrix Scientific

073848-250MG 073848-1G 073848-5G
102533-314EA 427.69 USD
102533-314 102533-316 102533-318
1-(Benzo[d]oxazol-2-yl)ethanone ≥95%
1-(Benzo[d]oxazol-2-yl)ethanone

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.1574 g/mol
MDL Number: MFCD00223823
CAS Number: 122433-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR