2-Methyl-1,3-benzoxazole-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

073847-250MG 073847-1G 073847-5G
102533-308EA 1018.46 USD
102533-308 102533-310 102533-312
2-Methyl-1,3-benzoxazole-6-carboxylic acid ≥95%
2-Methyl-1,3-benzoxazole-6-carboxylic acid

Hazard: Irritant

Formula: C₉H₇NO₃
MW: 177.1600 g/mol
MDL Number: MFCD07787266
CAS Number: 13452-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR