(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥95%

Supplier: Matrix Scientific

Synonyms: XANTPHOS, 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

073845-5G 073845-25G
102533-298EA 126.15 USD
102533-298 102833-654
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) ≥95%
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)

Hazard: Irritant

Formula: C₃₉H₃₂OP₂
MW: 578.62 g/mol
Melting Pt: 224…228 °C
MDL Number: MFCD00233866
CAS Number: 161265-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR